missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Hydroxyphenyl)ethanol, 98%

Manufacturer:  Alfa Aesar™ A1489114

 View more versions of this product

Catalog No. AAA1489114



Specifications

Specifications

2-(4-Hydroxyphenyl)ethanol
89°C to 92°C
501-94-0
MFCD00002902
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
C1=CC(=CC=C1CCO)O
138.166
CHEBI:1879
98%
139°C (1mmHg)
25g
C8H10O2
1859884
YCCILVSKPBXVIP-UHFFFAOYSA-N
4-(2-hydroxyethyl)phenol
10393
138.17

Chemical Identifiers

501-94-0
138.166
YCCILVSKPBXVIP-UHFFFAOYSA-N
10393
4-(2-hydroxyethyl)phenol
C8H10O2
MFCD00002902
tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
CHEBI:1879
C1=CC(=CC=C1CCO)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents