Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride
CAS 57591-61-4
Formula Weight 217.65
MDL Number MFCD01862152
Synonym (R)-Amino-(4-hydroxyphenyl)acetic acid methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6560103

  SDS
alfa aesar™
H6560103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6560106

  SDS
alfa aesar™
H6560106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6560114

  SDS
alfa aesar™
H6560114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 187° to 190°C
Formula Weight 217.65
Chemical Name or Material 2-(4-Hydroxyphenyl)-D-glycine methyl ester hydrochloride
CAS 57591-61-4
MDL Number MFCD01862152
Physical Form Solid
Assay 95%
Quantity 1g
Synonym (R)-Amino-(4-hydroxyphenyl)acetic acid methyl ester hydrochloride