Please login to your online account to display your discounted pricing

2-(4-Hydroxyphenoxy)propionic acid, 98%, ACROS Organics™

$85.87 - $85.87

Chemical Identifiers

Chemical Name or Material 2-(4-Hydroxyphenoxy)propionic acid
Name Note 98%
CAS 67648-61-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10O4
Linear Formula CH3CH(OC6H4OH)CO2H
Formula Weight 182.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC296150010

  SDS
thermo fisher scientific
296150010
Glass bottle 1g Each for $85.87
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 182.18
Physical Form Crystalline Powder
Quantity 1g
Color White to light beige
TSCA TSCA
Melting Point 143° to 147°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
CAS 67648-61-7
Molecular Formula C9H10O4
Linear Formula CH3CH(OC6H4OH)CO2H
MDL Number MFCD00012181
Name Note 98%
Assay Percent Range 98%
Chemical Name or Material 2-(4-Hydroxyphenoxy)propionic acid
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.