Learn More
2,4-Hexadienoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120531000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 4-Hexadienoic acid | |
110-44-1 | |
100.0 | |
Cream-White or White | |
Authentic | |
Plastic bottle | |
CH3CH=CHCH=CHCO2H | |
100 g | |
15,296 | |
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid | |
WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
(2E,4E)-hexa-2,4-dienoic acid | |
643460 | |
112.13 | |
Crystalline Powder |
99% | |
98.5 | |
132°C to 135°C | |
127°C | |
99% | |
C6H8O2 | |
MFCD00002703 | |
02, 483 | |
15, 8847 | |
Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in ethanol,chloroform,very soluble in diethyl ether | |
C\C=C\C=C\C(O)=O | |
112.13 | |
CHEBI:38358 | |
99% |
Chemical Identifiers
110-44-1 | |
112.13 | |
WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
643460 | |
(2E,4E)-hexa-2,4-dienoic acid |
C6H8O2 | |
MFCD00002703 | |
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid | |
CHEBI:38358 | |
C\C=C\C=C\C(O)=O |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-768-7
RTECSNumber : WG2100000
TSCA : TSCA
RUO – Research Use Only