missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Hexadiene, tech. 90%, mixture of isomers

Manufacturer:  Alfa Aesar™ H5340403

 View more versions of this product

Catalog No. AAH5340403



Description

Description

Applications
2,4-Hexadiene is used in the production of 3,6-dimethyl-3,6-dihydro-[1,2]dithiine.

Solubility
Miscible with ethanol, ethyl ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2,4-Hexadiene
82°C
mixture of isomers
C6H10
MFCD00009297
unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
CC=CC=CC
82.146
82.15
Technical
−14°C (−7°F)
1g
592-46-1
CH3CH=CHCH=CHCH3
UN2458
APPOKADJQUIAHP-CIIODKQPSA-N
(2Z,4E)-hexa-2,4-diene
643786
90%

Chemical Identifiers

592-46-1
82.146
APPOKADJQUIAHP-CIIODKQPSA-N
643786
CC=CC=CC
C6H10
MFCD00009297
unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
(2Z,4E)-hexa-2,4-diene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXADIENES

SDS Documents