missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Hexadienal, predominantly trans,trans, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1320014

 View more versions of this product

Catalog No. AAA1320014



Description

Description

2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.

Solubility
It is Insoluble in water, but soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Store under inert gas.Take measures to prevent the build up of electrostatic charge. Incompatible with strong reducing agents, strong bases, strong oxidizing agents. It is Stable, Combustible.
Specifications

Specifications

2,4-Hexadienal
142-83-6
170°C to 173°C
Floral
1.5386
25 g
1698401
2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein
BATOPAZDIZEVQF-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienal
637564
96.13
predominantly trans,trans
0.871
67°C (152°F)
C6H8O
MFCD00007004
UN2922
Air sensitive
It is Insoluble in water,but soluble in ethanol.
CC=CC=CC=O
96.129
CHEBI:82334
95%

Chemical Identifiers

142-83-6
96.129
BATOPAZDIZEVQF-MQQKCMAXSA-N
637564
(2E,4E)-hexa-2,4-dienal
C6H8O
MFCD00007004
2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein
CHEBI:82334
CC=CC=CC=O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H227-H317
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.

P201-P202-P210-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P361-P363-P370+P378q-P501c

H227-H302-H311-H315-H319-H351

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 205-564-3

RTECSNumber : WG1925000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only