missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Heptadien-1-ol

Manufacturer:  Alfa Aesar™ 1576506

 View more versions of this product

Catalog No. AA1576506



Specifications

Specifications

2,4-Heptadien-1-ol
5g
C7H12O
hepta-2,4-dien-1-ol, 2,4-heptadien-1-ol, e,e-2,4-heptadien-1-ol, 2e,4e-hepta-2,4-dien-1-ol, trans-2-trans-4-heptadien-1-ol, 2,4-heptadien-1-ol, 2e,4e, unii-9954pjf37o, 2e,4e-2,4-heptadien-1-ol, 2e,4z-2,4-heptadien-1-ol, 2,4-heptadien-1-ol, e,e
CCC=CC=CCO
112.172
112.17
63° (145°F)
33467-79-7
MFCD00014049
MDRZSADXFOPYOC-VNKDHWASSA-N
(2E,4E)-hepta-2,4-dien-1-ol
5367391
Liquid

Chemical Identifiers

33467-79-7
112.172
MDRZSADXFOPYOC-VNKDHWASSA-N
5367391
CCC=CC=CCO
C7H12O
MFCD00014049
hepta-2,4-dien-1-ol, 2,4-heptadien-1-ol, e,e-2,4-heptadien-1-ol, 2e,4e-hepta-2,4-dien-1-ol, trans-2-trans-4-heptadien-1-ol, 2,4-heptadien-1-ol, 2e,4e, unii-9954pjf37o, 2e,4e-2,4-heptadien-1-ol, 2e,4z-2,4-heptadien-1-ol, 2,4-heptadien-1-ol, e,e
(2E,4E)-hepta-2,4-dien-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents