missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Heptadien-1-ol

$65.73 - $235.51

Chemical Identifiers

CAS 33467-79-7
Formula Weight 112.17
MDL Number MFCD00014049
Chemical Name or Material 2,4-Heptadien-1-ol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1576506 SDS Alfa Aesar™
1576506
5g Each for $65.73
AA1576514 SDS Alfa Aesar™
1576514
25g Each for $235.51
Specifications Description & Specifications

Chemical Identifiers

33467-79-7
112.17
MFCD00014049
2,4-Heptadien-1-ol

Specifications

63° (145°F)
33467-79-7
112.17
2,4-Heptadien-1-ol
5g
Liquid
MFCD00014049

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA