Please login to your online account to display your discounted pricing

2-[(4-Fluorophenyl)sulfonyl]-N'-hydroxyethanimidamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-[(4-Fluorophenyl)sulfonyl]-N';-hydroxyethanimidamide
CAS 175203-76-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9FN2O3S
Linear Formula FC6H4SO2CH2C(=NOH)NH2
Formula Weight 232.23
MDL Number MFCD00205110
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM08532DA

  SDS
fisher scientific company
KM08532DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM08532EA

  SDS
fisher scientific company
KM08532EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.23
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-[(4-Fluorophenyl)sulfonyl]-N';-hydroxyethanimidamide
Assay Percent Range 97%
CAS 175203-76-6
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Linear Formula FC6H4SO2CH2C(=NOH)NH2
MDL Number MFCD00205110
Packaging Amber Glass Bottle
Molecular Formula C8H9FN2O3S
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.