Please login to your online account to display your discounted pricing

2-[(4-Fluorophenyl)sulfonyl]-N'-hydroxyethanimidamide, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM08532EA

 View more versions of this product

Catalog No. KM08532EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 232.23
Quantity 10g
Assay 97%
CAS Min % 97.0
MDL Number MFCD00205110
CAS 175203-76-6
Packaging Amber Glass Bottle
Chemical Name or Material 2-[(4-Fluorophenyl)sulfonyl]-N';-hydroxyethanimidamide
Linear Formula FC6H4SO2CH2C(=NOH)NH2
Assay Percent Range 97%
Molecular Formula C8H9FN2O3S
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-[(4-Fluorophenyl)sulfonyl]-N';-hydroxyethanimidamide
CAS 175203-76-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9FN2O3S
Linear Formula FC6H4SO2CH2C(=NOH)NH2
Formula Weight 232.23
MDL Number MFCD00205110
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents