Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol, 95%

$48.36 - $188.35

Chemical Identifiers

Chemical Name or Material 2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol
CAS 853955-70-1
Molecular Formula C15H17FOSi
Formula Weight 260.38
MDL Number MFCD16251550
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5173503

  SDS
alfa aesar™
H5173503
1g Each for $48.36

AAH5173506

  SDS
alfa aesar™
H5173506
5g Each for $188.35
Description & Specifications

Specifications

Formula Weight 260.38
MDL Number MFCD16251550
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C15H17FOSi
Chemical Name or Material 2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol
Quantity 1g
CAS 853955-70-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.