Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Fluorophenyl)acetamidoxime, 97%

Chemical Identifiers

Chemical Name or Material 2-(4-Fluorophenyl)acetamidoxime
Formula Weight 168.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6699903

 
alfa aesar™
H6699903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66999MD

 
alfa aesar™
H66999MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.17
Physical Form Solid
Assay 97%
Chemical Name or Material 2-(4-Fluorophenyl)acetamidoxime
Color White
Quantity 1g