Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Fluorophenyl)-2-methylmorpholine, 99%

$119.45 - $415.61

Chemical Identifiers

Chemical Name or Material 2-(4-Fluorophenyl)-2-methylmorpholine
CAS 109461-46-3
Molecular Formula C11H14FNO
Formula Weight 195.24
MDL Number MFCD08061112
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5100703

 
alfa aesar™
H5100703
1g Each for $415.61

AAH51007MD

 
alfa aesar™
H51007MD
250mg Each for $119.45
Description & Specifications

Specifications

Formula Weight 195.24
Chemical Name or Material 2-(4-Fluorophenyl)-2-methylmorpholine
Assay 99%
Molecular Formula C11H14FNO
MDL Number MFCD08061112
CAS 109461-46-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.