Please login to your online account to display your discounted pricing

2-(4-Fluorophenoxy)nicotinic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00878EA

 View more versions of this product

Catalog No. TL00878EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 233.2
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-(4-Fluorophenoxy)nicotinic acid
Assay Percent Range 97%
Molecular Formula C12H8FNO3
MDL Number MFCD00833409
Packaging Amber Glass Bottle
CAS 54629-13-9

Specifications

Chemical Name or Material 2-(4-Fluorophenoxy)nicotinic acid
CAS 54629-13-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H8FNO3
Formula Weight 233.2
MDL Number MFCD00833409