Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Fluorophenoxy)nicotinic acid, 97+%

$50.18 - $181.64

Chemical Identifiers

Chemical Name or Material 2-(4-Fluorophenoxy)nicotinic acid
CAS 54629-13-9
Molecular Formula C12H8FNO3
Formula Weight 233.2
MDL Number MFCD00833409
Synonym 2-(4-Fluorophenoxy)pyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2153303

 
alfa aesar™
B2153303
1g Each for $50.18

AAB2153306

 
alfa aesar™
B2153306
5g Each for $181.64
Description & Specifications

Specifications

Formula Weight 233.2
Chemical Name or Material 2-(4-Fluorophenoxy)nicotinic acid
Assay ≥97%
Molecular Formula C12H8FNO3
MDL Number MFCD00833409
CAS 54629-13-9
Synonym 2-(4-Fluorophenoxy)pyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 182° to 185°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.