Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Fluorophenoxy)-N-methylbenzylamine, 96%

$83.93 - $304.36

Chemical Identifiers

Chemical Name or Material 2-(4-Fluorophenoxy)-N-methylbenzylamine
CAS 833482-53-4
Molecular Formula C14H14FNO
Formula Weight 231.27
MDL Number MFCD08064245
Synonym 1-[2-(4-Fluorophenoxy)phenyl]-N-methylmethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5075503

  SDS
alfa aesar™
H5075503
1g Each for $304.36

AAH50755MD

  SDS
alfa aesar™
H50755MD
250mg Each for $83.93
Description & Specifications

Specifications

Formula Weight 231.27
Chemical Name or Material 2-(4-Fluorophenoxy)-N-methylbenzylamine
Assay 96%
Molecular Formula C14H14FNO
MDL Number MFCD08064245
CAS 833482-53-4
Synonym 1-[2-(4-Fluorophenoxy)phenyl]-N-methylmethylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
2-(4-Fluorophenoxy)-N-methylbenzylamine is used as pharmaceutical-intermediates.

Notes
Air Sensitive. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents and air. Store under dry inert gas.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.