Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[4-Fluoro-3-(trifluoromethyl)phenyl]ethanol, 90+%

$56.50 - $187.80

Chemical Identifiers

Chemical Name or Material 2-[4-Fluoro-3-(trifluoromethyl)phenyl]ethanol
CAS 886761-80-4
Molecular Formula C9H8F4O
Formula Weight 208.15
MDL Number MFCD06660359
Synonym 4-Fluoro-3-(trifluoromethyl)phenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3286703

  SDS
alfa aesar™
H3286703
1g Each for $187.80

AAH32867MD

  SDS
alfa aesar™
H32867MD
250mg Each for $56.50
Description & Specifications

Specifications

Formula Weight 208.15
Chemical Name or Material 2-[4-Fluoro-3-(trifluoromethyl)phenyl]ethanol
Assay ≥90%
Molecular Formula C9H8F4O
MDL Number MFCD06660359
CAS 886761-80-4
Synonym 4-Fluoro-3-(trifluoromethyl)phenethyl alcohol
Quantity 1g