missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Ethylphenyl)thiazole-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5437406

 View more versions of this product

Catalog No. AAH5437406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(4-Ethylphenyl)thiazole-4-carboxylic acid
847744-15-4
MFCD07324809
OKIXVEUDAQRHLQ-UHFFFAOYSA-N
2-(4-ethylphenyl)-1,3-thiazole-4-carboxylic acid
4962607
97%
5g
C12H11NO2S
2-4-ethylphenyl thiazole-4-carboxylic acid
CCC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
233.285
233.28

Chemical Identifiers

847744-15-4
233.285
OKIXVEUDAQRHLQ-UHFFFAOYSA-N
4962607
CCC1=CC=C(C=C1)C2=NC(=CS2)C(=O)O
C12H11NO2S
MFCD07324809
2-4-ethylphenyl thiazole-4-carboxylic acid
2-(4-ethylphenyl)-1,3-thiazole-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents