Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Ethylphenyl)-5-n-propylpyrimidine, 99+%

$41.58 - $221.50

Chemical Identifiers

Chemical Name or Material 2-(4-Ethylphenyl)-5-n-propylpyrimidine
CAS 98495-11-5
Molecular Formula C15H18N2
Formula Weight 226.32
MDL Number MFCD00799380
Synonym 5-n-Propyl-2-pyrimidinethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2736103

  SDS
alfa aesar™
H2736103
1g Each for $41.58

AAH2736109

  SDS
alfa aesar™
H2736109
10g Each for $221.50
Description & Specifications

Specifications

Formula Weight 226.32
Synonym 5-n-Propyl-2-pyrimidinethiol
Molecular Formula C15H18N2
Chemical Name or Material 2-(4-Ethylphenyl)-5-n-propylpyrimidine
MDL Number MFCD00799380
CAS 98495-11-5
Assay ≥99%
Melting Point 41° to 43°C
Quantity 1g