Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-([4-(Ethoxycarbonyl)phenyl]dimethylsilyl)benzyl alcohol, 95%

$68.19 - $283.23

Chemical Identifiers

Chemical Name or Material 2-([4-(Ethoxycarbonyl)phenyl]dimethylsilyl)benzyl alcohol
CAS 1244855-68-2
Molecular Formula C18H22O3Si
Formula Weight 314.46
MDL Number MFCD16251591
Synonym Ethyl 4-[(2-hydroxymethylphenyl)dimethylsilyl]benzoate; 4-[(2-Hydroxymethylphenyl)dimethylsilyl]benzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5193203

 
alfa aesar™
H5193203
1g Each for $68.19

AAH5193206

 
alfa aesar™
H5193206
5g Each for $283.23
Description & Specifications

Specifications

Formula Weight 314.46
Chemical Name or Material 2-([4-(Ethoxycarbonyl)phenyl]dimethylsilyl)benzyl alcohol
Assay 95%
Molecular Formula C18H22O3Si
MDL Number MFCD16251591
CAS 1244855-68-2
Synonym Ethyl 4-[(2-hydroxymethylphenyl)dimethylsilyl]benzoate; 4-[(2-Hydroxymethylphenyl)dimethylsilyl]benzoic acid ethyl ester
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.