missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Diphenyl-1,3-thiazole-5-sulfonyl chloride, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 868755-57-1
Molecular Formula C15H10ClNO2S2
Molecular Weight (g/mol) 335.82
MDL Number MFCD08271911
InChI Key AWNJOBYLGNWNCQ-UHFFFAOYSA-N
Synonym diphenyl-1,3-thiazole-5-sulfonyl chloride, 2,4-diphenylthiazole-5-sulfonyl chloride, 5-thiazolesulfonylchloride, 2,4-diphenyl, 5-chlorosulphonyl-2,4-diphenyl-1,3-thiazole
PubChem CID 18525757
IUPAC Name diphenyl-1,3-thiazole-5-sulfonyl chloride
SMILES ClS(=O)(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC42303CB
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC42303CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

868755-57-1
335.82
AWNJOBYLGNWNCQ-UHFFFAOYSA-N
18525757
ClS(=O)(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
C15H10ClNO2S2
MFCD08271911
diphenyl-1,3-thiazole-5-sulfonyl chloride, 2,4-diphenylthiazole-5-sulfonyl chloride, 5-thiazolesulfonylchloride, 2,4-diphenyl, 5-chlorosulphonyl-2,4-diphenyl-1,3-thiazole
diphenyl-1,3-thiazole-5-sulfonyl chloride

Specifications

868755-57-1
100.0
Amber Glass Bottle
MFCD08271911
diphenyl-1,3-thiazole-5-sulfonyl chloride, 2,4-diphenylthiazole-5-sulfonyl chloride, 5-thiazolesulfonylchloride, 2,4-diphenyl, 5-chlorosulphonyl-2,4-diphenyl-1,3-thiazole
ClS(=O)(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
335.82
335.83
2, 4-Diphenyl-1, 3-thiazole-5-sulfonyl chloride
96.5
97%
C15H10ClNO2S2
250mg
AWNJOBYLGNWNCQ-UHFFFAOYSA-N
diphenyl-1,3-thiazole-5-sulfonyl chloride
18525757
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'