missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2,4-Diphenyl-1,3-thiazol-5-yl)methylamine hydrochloride, ≥95%, Maybridge

Chemical Identifiers

CAS 879896-44-3
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C16H14N2S·ClH
Formula Weight 302.83
MDL Number MFCD08435866
Chemical Name or Material (2, 4-Diphenyl-1, 3-thiazol-5-yl)methylamine hydrochloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC42363DA SDS Fisher Scientific Company
CC42363DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

879896-44-3
100.0
C16H14N2S·ClH
MFCD08435866
95%
94.5
95%
302.83
(2, 4-Diphenyl-1, 3-thiazol-5-yl)methylamine hydrochloride

Specifications

1g
94.5
95%
302.83
(2, 4-Diphenyl-1, 3-thiazol-5-yl)methylamine hydrochloride
879896-44-3
100.0
C16H14N2S·ClH
MFCD08435866
95%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.