missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ H5483306

 View more versions of this product

Catalog No. AAH5483306



Specifications

Specifications

2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride
56044-12-3
MFCD07186482
2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfony
C1=CC2=C(C=C1S(=O)(=O)Cl)C(=O)NC(=O)N2
260.648
259.97
5g
C8H5ClN2O4S
UN3261
LEFXQSDJQGOMPS-UHFFFAOYSA-N
2,4-dioxo-1H-quinazoline-6-sulfonyl chloride
3162025
97%

Chemical Identifiers

56044-12-3
260.648
LEFXQSDJQGOMPS-UHFFFAOYSA-N
3162025
C1=CC2=C(C=C1S(=O)(=O)Cl)C(=O)NC(=O)N2
C8H5ClN2O4S
MFCD07186482
2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfony
2,4-dioxo-1H-quinazoline-6-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents