missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ H54833MD

 View more versions of this product

Catalog No. AAH54833MD



Specifications

Specifications

2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride
250mg
C8H5ClN2O4S
UN3261
LEFXQSDJQGOMPS-UHFFFAOYSA-N
2,4-dioxo-1H-quinazoline-6-sulfonyl chloride
3162025
97%
Moisture sensitive
56044-12-3
MFCD07186482
2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfony
C1=CC2=C(C=C1S(=O)(=O)Cl)C(=O)NC(=O)N2
260.648
259.97

Chemical Identifiers

56044-12-3
260.648
LEFXQSDJQGOMPS-UHFFFAOYSA-N
3162025
C1=CC2=C(C=C1S(=O)(=O)Cl)C(=O)NC(=O)N2
C8H5ClN2O4S
MFCD07186482
2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfony
2,4-dioxo-1H-quinazoline-6-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents