missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H54410MD

 View more versions of this product

Catalog No. AAH54410MD



Description

Description

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications

Specifications

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
C4H3ClN2O4S
250 mg
Moisture sensitive
FQHNJCWWBKYCJW-UHFFFAOYSA-N
2,4-dioxo-1H-pyrimidine-5-sulfonyl chloride
234620
97%
28485-18-9
MFCD06655485
UN3261
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride, 1,2,3,4-tetrahydro-2,4-dioxo-5-pyrimidinesulfonyl chloride, 5-pyrimidinesulfonylchloride, 1,2,3,4-tetrahydro-2,4-dioxo, uracil-5-sulphonyl chloride, 5-chlorosulfonyl-1,3-dihydropyrimidine-2,4-dione, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinesulfonyl chloride, 2,4-dioxo-1,2,3,4-tetrahydro-pyrimidine-5-sulfonyl chloride
ClS(=O)(=O)C1=CNC(=O)NC1=O
210.59
209.95

Chemical Identifiers

28485-18-9
210.59
FQHNJCWWBKYCJW-UHFFFAOYSA-N
234620
C4H3ClN2O4S
MFCD06655485
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride, 2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride, 1,2,3,4-tetrahydro-2,4-dioxo-5-pyrimidinesulfonyl chloride, 5-pyrimidinesulfonylchloride, 1,2,3,4-tetrahydro-2,4-dioxo, uracil-5-sulphonyl chloride, 5-chlorosulfonyl-1,3-dihydropyrimidine-2,4-dione, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinesulfonyl chloride, 2,4-dioxo-1,2,3,4-tetrahydro-pyrimidine-5-sulfonyl chloride
ClS(=O)(=O)C1=CNC(=O)NC1=O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only