missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dinitrophenylhydrazine, Spectrum™ Chemical
DI149, 119-26-6, C6H6N4O4
$157.89 - $41732.69
Chemical Identifiers
CAS | 119-26-6 |
---|---|
Molecular Formula | C6H6N4O4 |
Molecular Weight (g/mol) | 198.14 |
InChI Key | HORQAOAYAYGIBM-UHFFFAOYSA-N |
IUPAC Name | (2,4-dinitrophenyl)hydrazine |
SMILES | NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18600085
|
Spectrum Chemical Mfg Cor
DI14925GM |
25 g | Amber Glass Bottle |
Each for $157.89
|
|
||||
18600089
|
Spectrum Chemical Mfg Cor
DI149500GM |
500 g | Amber Glass Bottle |
Each for $1,459.57
|
|
||||
18600088
|
Spectrum Chemical Mfg Cor
DI14910KGBL |
10 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18600087
|
Spectrum Chemical Mfg Cor
DI14950KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical 2,4-Dinitrophenylhydrazine is a chemical compound that is usually supplied as a wet red-to-orange solid that is shock sensitive. It is used in Brady′s test to detect the carbonyl functionality of an aldehyde or ketone functional group.Chemical Identifiers
119-26-6 | |
198.14 | |
(2,4-dinitrophenyl)hydrazine |
C6H6N4O4 | |
HORQAOAYAYGIBM-UHFFFAOYSA-N | |
NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O |
Specifications
119-26-6 | |
Amber Glass Bottle | |
25 g | |
HORQAOAYAYGIBM-UHFFFAOYSA-N | |
(2,4-dinitrophenyl)hydrazine | |
Reagent |
100% | |
C6H6N4O4 | |
UN1325 | |
NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O | |
198.14 |