Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dinitrophenyl thiocyanate, 98%

$138.54 - $455.62

Chemical Identifiers

Chemical Name or Material 2,4-Dinitrophenyl thiocyanate
CAS 1594-56-5
Molecular Formula C7H3N3O4S
Formula Weight 225.18
MDL Number MFCD00047702
Beilstein 1983292
Synonym 2,4-Dinitrothiocyanatobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2414714

 
alfa aesar™
B2414714
25g Each for $138.54

AAB2414722

 
alfa aesar™
B2414722
100g Each for $455.62
Description & Specifications

Specifications

Formula Weight 225.18
Chemical Name or Material 2,4-Dinitrophenyl thiocyanate
Assay 98%
Molecular Formula C7H3N3O4S
MDL Number MFCD00047702
CAS 1594-56-5
Beilstein 1983292
Synonym 2,4-Dinitrothiocyanatobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XK9450000
Odor Irritating
Melting Point 138° to 140°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.