missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dinitrobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1453606

 View more versions of this product

Catalog No. AAA1453606



Description

Description

Applications
2,4-Dinitrobenzoic acid was used in the spectrophotometric determination of diazepam in pure samples and in its pharmaceutical preparations. It was employed as chromophore probe for analysis of perfluorinated carboxylic acids in water by capillary zone electrophoresis. It can be used to produce 2,4-dinitro-benzoyl chloride.

Solubility
Soluble in dioxane 50 mg/ml.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2,4-Dinitrobenzoic acid
5g
C7H4N2O6
658650
ZIIGSRYPZWDGBT-UHFFFAOYSA-N
212.117
212.12
178°C to 184°C
610-30-0
MFCD00017006
2,4-dinitrobenzoic acid, benzoic acid, 2,4-dinitro, ccris 1766, acmc-20aoc2, 2.4-dinitrobenzoic acid, benzoicacid, 2,4-dinitro, 2,4-dinitrobenzoic acidanion, 4-09-00-01239 beilstein handbook reference, aronis26960, 1-carboxy-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)O
11880
98%

Chemical Identifiers

610-30-0
212.117
ZIIGSRYPZWDGBT-UHFFFAOYSA-N
11880
C7H4N2O6
MFCD00017006
2,4-dinitrobenzoic acid, benzoic acid, 2,4-dinitro, ccris 1766, acmc-20aoc2, 2.4-dinitrobenzoic acid, benzoicacid, 2,4-dinitro, 2,4-dinitrobenzoic acidanion, 4-09-00-01239 beilstein handbook reference, aronis26960, 1-carboxy-2,4-dinitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG9140500

TSCA : TSCA

Documents