Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dinitroanisole, 98%

$64.55 - $64.55

Chemical Identifiers

Chemical Name or Material 2,4-Dinitroanisole
CAS 119-27-7
Molecular Formula C7H6N2O5
Formula Weight 198.14g/mol
MDL Number MFCD00035745
UN Number UN2811
Beilstein 1881474
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2154214

 
alfa aesar™
B21542-14
25g Each for $64.55
Description & Specifications

Specifications

Melting Point 88° to 94°C
Boiling Point 207°C (12mmHg)
Quantity 25g
Chemical Name or Material 2,4-Dinitroanisole
CAS 119-27-7
Assay 98%
Molecular Formula C7H6N2O5
Formula Weight 198.14g/mol
MDL Number MFCD00035745
UN Number UN2811
Beilstein 1881474
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number DA5250000
TSCA TSCA

Applications
It can be used as a dye ingredient. It is one of the insensitive nitroaromatic ingredients increasingly used as a replacement for 2,4,6-trinitrotoluene (TNT) in munitions.

Solubility
Solubility in hot methanol; almost transparency. Water solubility, 155 mg/L 15°C. Completely soluble in ethyl acetate & acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.