Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dinitro-1-naphthol, 95%

$27.08 - $324.78

Chemical Identifiers

Chemical Name or Material 2,4-Dinitro-1-naphthol
CAS 605-69-6
Molecular Formula C10H6N2O5
Formula Weight 234.17
MDL Number MFCD00065352
Beilstein 2057462
Synonym C.I. 10315; Martius Yellow
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2557006

 
alfa aesar™
B2557006
5g Each for $27.08

AAB2557014

 
alfa aesar™
B2557014
25g Each for $94.04

AAB2557022

 
alfa aesar™
B2557022
100g Each for $324.78
Description & Specifications

Specifications

Formula Weight 234.17
Chemical Name or Material 2,4-Dinitro-1-naphthol
TSCA TSCA
Assay 95%
Molecular Formula C10H6N2O5
MDL Number MFCD00065352
CAS 605-69-6
Beilstein 2057462
Synonym C.I. 10315; Martius Yellow
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QL3850000
Melting Point 140°C (dec.)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.