missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylthiazole-5-sulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ H5457806

 View more versions of this product

Catalog No. AAH5457806



Description

Description

Specifications

Specifications

2,4-Dimethylthiazole-5-sulfonyl chloride
82°C to 84°C (0.3mmHg)
80466-80-4
MFCD00052746
2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, 2,4-dimethylthiazol-5-ylsulphonyl chloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 5-thiazolesulfonylchloride, 2,4-dimethyl, dimethyl-1,3-thiazole-5-sulfonyl chloride
CC1=C(SC(=N1)C)S(=O)(=O)Cl
211.678
211.68
Moisture sensitive
5g
C5H6ClNO2S2
UN3265
GFFJSTHQILQFNQ-UHFFFAOYSA-N
2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride
2776248
97%

Chemical Identifiers

80466-80-4
211.678
GFFJSTHQILQFNQ-UHFFFAOYSA-N
2776248
CC1=C(SC(=N1)C)S(=O)(=O)Cl
C5H6ClNO2S2
MFCD00052746
2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, 2,4-dimethylthiazol-5-ylsulphonyl chloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 5-thiazolesulfonylchloride, 2,4-dimethyl, dimethyl-1,3-thiazole-5-sulfonyl chloride
2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents