Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylphenylurea, 98%

$26.23 - $92.36

Chemical Identifiers

Chemical Name or Material 2,4-Dimethylphenylurea
CAS 2990-02-5
Molecular Formula C9H12N2O
Formula Weight 164.2
MDL Number MFCD00025409
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2566806

  SDS
alfa aesar™
B2566806
5g Each for $26.23

AAB2566814

  SDS
alfa aesar™
B2566814
25g Each for $92.36
Description & Specifications

Specifications

Formula Weight 164.2
CAS 2990-02-5
Chemical Name or Material 2,4-Dimethylphenylurea
Molecular Formula C9H12N2O
Quantity 5g
Assay 98%
Melting Point 203° to 204°C (dec.)
MDL Number MFCD00025409