missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylphenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2100914

 View more versions of this product

Catalog No. AAB2100914



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dimethylphenylhydrazine hydrochloride
∼188°C (decomposition)
MFCD00013381
Hygroscopic
ZLGXGLXFBMIVPU-UHFFFAOYSA-N
(2,4-dimethylphenyl)hydrazine;hydrochloride
16213059
97%
60480-83-3
C8H13ClN2
25 g
2,4-dimethylphenylhydrazine hydrochloride, 2,4-dimethylphenyl hydrazine hydrochloride, 1-2,4-dimethylphenyl hydrazine hydrochloride, 2,4-dimethylphenylhydrazinehydrochloride, 2,4-dimethylphenyldrazine hydrochloride, hydrazine, 2,4-dimethylphenyl-, monohydrochloride, pubchem6399, acmc-1bh4y, ksc491e7t, 2,4-xylylhydrazine hydrochloride
CC1=CC(=C(C=C1)NN)C.Cl
172.656
172.66

Chemical Identifiers

60480-83-3
172.656
ZLGXGLXFBMIVPU-UHFFFAOYSA-N
16213059
CC1=CC(=C(C=C1)NN)C.Cl
C8H13ClN2
MFCD00013381
2,4-dimethylphenylhydrazine hydrochloride, 2,4-dimethylphenyl hydrazine hydrochloride, 1-2,4-dimethylphenyl hydrazine hydrochloride, 2,4-dimethylphenylhydrazinehydrochloride, 2,4-dimethylphenyldrazine hydrochloride, hydrazine, 2,4-dimethylphenyl-, monohydrochloride, pubchem6399, acmc-1bh4y, ksc491e7t, 2,4-xylylhydrazine hydrochloride
(2,4-dimethylphenyl)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only