Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylphenylacetic acid, 98%

$102.31 - $354.09

Chemical Identifiers

Chemical Name or Material 2,4-Dimethylphenylacetic acid
CAS 6331-04-0
Molecular Formula C10H12O2
Formula Weight 164.21
MDL Number MFCD00014369
Synonym (2,4-Xylyl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5028403

  SDS
alfa aesar™
H5028403
1g Each for $354.09

AAH50284MD

  SDS
alfa aesar™
H50284MD
250mg Each for $102.31
Description & Specifications

Specifications

Formula Weight 164.21
Chemical Name or Material 2,4-Dimethylphenylacetic acid
Assay 98%
Molecular Formula C10H12O2
MDL Number MFCD00014369
CAS 6331-04-0
Quantity 1g
Synonym (2,4-Xylyl)acetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.