missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylphenyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1166514

 View more versions of this product

Catalog No. AAL1166514



Specifications

Specifications

2,4-Dimethylphenyl isothiocyanate
25°C to 29°C
39842-01-8
MFCD00022054
2690174
HOHSBFCSOARUBF-UHFFFAOYSA-N
1-isothiocyanato-2,4-dimethylbenzene
142386
98%
122°C to 130°C (8mmHg)
25g
C9H9NS
UN2923
2,4-dimethylphenyl isothiocyanate, 2,4-dimethylphenylisothiocyanate, 2,4-xylyl isothiocyanate, 1-isothiocyanato-2,4-dimethyl-benzene, 2,4-dimethylbenzenisothiocyanate, xylyl isothiocyanate, 4-isothiocyanato-m-xylene, acmc-1agr7, 2,4-dimethylphenyl-isothiocyanate, 2,4-dimethyl-phenyl isothiocyanate
CC1=CC(=C(C=C1)N=C=S)C
163.238
163.24

Chemical Identifiers

39842-01-8
163.238
HOHSBFCSOARUBF-UHFFFAOYSA-N
142386
CC1=CC(=C(C=C1)N=C=S)C
C9H9NS
MFCD00022054
2,4-dimethylphenyl isothiocyanate, 2,4-dimethylphenylisothiocyanate, 2,4-xylyl isothiocyanate, 1-isothiocyanato-2,4-dimethyl-benzene, 2,4-dimethylbenzenisothiocyanate, xylyl isothiocyanate, 4-isothiocyanato-m-xylene, acmc-1agr7, 2,4-dimethylphenyl-isothiocyanate, 2,4-dimethyl-phenyl isothiocyanate
1-isothiocyanato-2,4-dimethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents