Learn More
2,4-Dimethylphenyl isocyanate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1112603
Description
2,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dimethylphenyl isocyanate is used as a pharmaceutical intermediate.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture, heat, alcohols and amines.
Specifications
2,4-Dimethylphenyl isocyanate | |
1.041 | |
1 g | |
C9H9NO | |
UN2206 | |
2,4-dimethylphenyl isocyanate, 2,4-dimethylphenylisocyanate, benzene, 1-isocyanato-2,4-dimethyl, 2,4-dimethylbenzenisocyanate, acmc-20aoc8, 1-isocyanato-2,4-dimethyl-benzene | |
QUOBVYPFBJUOAJ-UHFFFAOYSA-N | |
1-isocyanato-2,4-dimethylbenzene | |
5207585 | |
≥98% |
51163-29-2 | |
114°C to 115°C (2 mmHg) | |
1.535 | |
MFCD00013853 | |
386756 | |
Hydrolyzes in water. | |
CC1=CC(=C(C=C1)N=C=O)C | |
147.177 | |
147.18 |
Chemical Identifiers
51163-29-2 | |
147.177 | |
QUOBVYPFBJUOAJ-UHFFFAOYSA-N | |
5207585 | |
CC1=CC(=C(C=C1)N=C=O)C |
C9H9NO | |
MFCD00013853 | |
2,4-dimethylphenyl isocyanate, 2,4-dimethylphenylisocyanate, benzene, 1-isocyanato-2,4-dimethyl, 2,4-dimethylbenzenisocyanate, acmc-20aoc8, 1-isocyanato-2,4-dimethyl-benzene | |
1-isocyanato-2,4-dimethylbenzene |
Safety and Handling
GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P260-P264b-P270-P271-P272-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H301-H315-H317-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only