missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylphenoxyacetic acid, 98+%

$58.92 - $189.68

Chemical Identifiers

CAS 13334-49-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00014355
InChI Key XRTZHWXWLUGOAT-UHFFFAOYSA-N
Synonym 2-2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxyacetic acid, 2,4-xylyloxyacetic acid, 2-2,4-dimethylphenoxy ethanoic acid, 2,4-dimethylphenoxyaceticacid, akos bbb/200, akos bc-2696, acmc-1btqb, labotest-bb lt00847868
PubChem CID 83354
IUPAC Name 2-(2,4-dimethylphenoxy)acetic acid
SMILES CC1=CC(=C(C=C1)OCC(=O)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0129606 View Documents Alfa Aesar™
L0129606
5g Each for $58.92
 
AAL0129614 View Documents Alfa Aesar™
L0129614
25g Each for $189.68
 
Specifications

Chemical Identifiers

13334-49-1
180.203
XRTZHWXWLUGOAT-UHFFFAOYSA-N
83354
CC1=CC(=C(C=C1)OCC(=O)O)C
C10H12O3
MFCD00014355
2-2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxyacetic acid, 2,4-xylyloxyacetic acid, 2-2,4-dimethylphenoxy ethanoic acid, 2,4-dimethylphenoxyaceticacid, akos bbb/200, akos bc-2696, acmc-1btqb, labotest-bb lt00847868
2-(2,4-dimethylphenoxy)acetic acid

Specifications

139°C to 143°C
13334-49-1
MFCD00014355
2-2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxyacetic acid, 2,4-xylyloxyacetic acid, 2-2,4-dimethylphenoxy ethanoic acid, 2,4-dimethylphenoxyaceticacid, akos bbb/200, akos bc-2696, acmc-1btqb, labotest-bb lt00847868
CC1=CC(=C(C=C1)OCC(=O)O)C
180.203
180.2
2,4-Dimethylphenoxyacetic acid
5g
C10H12O3
2263832
XRTZHWXWLUGOAT-UHFFFAOYSA-N
2-(2,4-dimethylphenoxy)acetic acid
83354
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning