missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylphenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC408455000

 View more versions of this product

Catalog No. AC408455000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 4-Dimethylphenol
Clear colorless to yellow
105-67-9
Authentic
Glass bottle
Liquid
99%
(CH3)2C6H3OH
MFCD00002233
15, 10279
1.027g/mL
500g
98.5
100.0
1.5375 to 1.5405 (20°C, 589nm)
≥98.5%
C8H10O
122.17
06, 486
2, 4-Xylenol

Chemical Identifiers

105-67-9
100.0
C8H10O
122.17
2, 4-Xylenol
≥98.5%
98.5
99%
(CH3)2C6H3OH
MFCD00002233
2, 4-Dimethylphenol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger