Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylphenol, 98%

$52.34 - $52.34

Chemical Identifiers

Chemical Name or Material 2,4-Dimethylphenol
CAS 105-67-9
Molecular Formula C8H10O
Formula Weight 122.17
MDL Number MFCD00002233
UN Number UN2261
Beilstein 636244
Merck Index 14,10082
Synonym 2,4-Xylenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1196622

  SDS
alfa aesar™
A1196622*
100g Each for $52.34
Description & Specifications

Specifications

Formula Weight 122.17
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,4-Dimethylphenol
TSCA TSCA
Assay 98%
Molecular Formula C8H10O
MDL Number MFCD00002233
CAS 105-67-9
UN Number UN2261
Beilstein 636244
Merck Index 14,10082
Synonym 2,4-Xylenol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
RTECS Number ZE5600000
Flash Point 94°C (201°F)
Boiling Point 210° to 212°C
Melting Point 21° to 23°C
Quantity 100g

Applications
It is used as a perfuming agent in cosmetic industry. Also used in the production of high-viscosity phosphate esters, as a feedstock for hindered phenol antioxidant and specialty modified phenolic resin manufacture. Also used as disinfectant and sanitizer on hospital premises.

Solubility
Slightly soluble in water 4068 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Store under nitrogen.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.