missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylimidazole, 97%

Manufacturer:  Alfa Aesar™ A1194906

 View more versions of this product

Catalog No. AAA1194906


 

Description

Description

Applications
It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications

Specifications

2,4-Dimethylimidazole
84°C to 88°C
930-62-1
C5H8N2
UN3263
LLPKQRMDOFYSGZ-UHFFFAOYSA-N
2,5-dimethyl-1H-imidazole
70259
266°C (733mmHg)
5g
97%
MFCD00022365
1639
CC1=CN=C(N1)C
96.133
96.13

Chemical Identifiers

930-62-1
96.133
LLPKQRMDOFYSGZ-UHFFFAOYSA-N
C5H8N2
MFCD00022365
70259
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.

RTECSNumber : NI4839712

SDS Documents