missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylimidazole, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1194914

 View more versions of this product

Catalog No. AAA1194914



Description

Description

It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a curing agent of epoxy resins. It is also widely used in the bonding, coating, capsulation of epoxy resins, or used for composite materials. It is also employed as medical raw material or used in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications

Specifications

2,4-Dimethylimidazole
84°C to 88°C
C5H8N2
25 g
1639
Soluble in water.
CC1=CN=C(C)N1
96.13
96.13
930-62-1
266°C (733 mmHg)
MFCD00022365
UN3263
2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
LLPKQRMDOFYSGZ-UHFFFAOYSA-N
2,5-dimethyl-1H-imidazole
70259
97%

Chemical Identifiers

930-62-1
96.13
LLPKQRMDOFYSGZ-UHFFFAOYSA-N
70259
CC1=CN=C(C)N1
C5H8N2
MFCD00022365
2,4-dimethylimidazole, 2,4-dimethyl-1h-imidazole, 1h-imidazole, 2,4-dimethyl, 2,4-dimethyl-3h-imidazole, 2,4-dimethyl-1h-imidazol, imidazole, 2,4-dimethyl, 2,4-dimethyl, 2,5-dimethylimidazole, imidazole,4-dimethyl, pubchem18587
2,5-dimethyl-1H-imidazole
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.

EINECSNumber : 213-221-4

RTECSNumber : NI4839712

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only