Please login to your online account to display your discounted pricing

2,4-Dimethylbenzylamine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365530050

 View more versions of this product

Catalog No. AC365530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 135.21
Quantity 5g
Color Clear light yellow
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C9H13N
Linear Formula (CH3)C6H3CHNH
CAS 94-98-4
CAS Min % 97.5
Health Hazard 1 Danger
Refractive Index 1.5360 to 1.5390 (20°C, 589nm)
Chemical Name or Material 2, 4-Dimethylbenzylamine
MDL Number MFCD00025575
Synonym 2, 4-Dimethylbenzenemethanamine
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 4-Dimethylbenzylamine
CAS 94-98-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H13N
Linear Formula (CH3)C6H3CHNH
Formula Weight 135.21
MDL Number MFCD00025575
Synonym 2, 4-Dimethylbenzenemethanamine
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents