Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylbenzyl chloride, 96%

Chemical Identifiers

Chemical Name or Material 2,4-Dimethylbenzyl chloride
CAS 824-55-5
Molecular Formula C9H11Cl
Formula Weight 154.64
MDL Number MFCD00015556
UN Number UN3265
Beilstein 742366
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0281006

  SDS
alfa aesar™
L0281006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0281014

  SDS
alfa aesar™
L0281014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 154.64
MDL Number MFCD00015556
Chemical Name or Material 2,4-Dimethylbenzyl chloride
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 1 Danger
CAS 824-55-5
UN Number UN3265
Assay 96%
Odor Pungent
Molecular Formula C9H11Cl
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 5g
Flash Point 96°C (204°F)
Beilstein 742366
TSCA TSCA
Boiling Point 103°C (15mmHg)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.