Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylbenzyl alcohol, 97%

$50.78 - $50.78

Chemical Identifiers

Chemical Name or Material 2,4-Dimethylbenzyl alcohol
CAS 16308-92-2
Molecular Formula C9H12O
Formula Weight 136.19
MDL Number MFCD00004624
Beilstein 1859679
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0289903

  SDS
alfa aesar™
L0289903
1g Each for $50.78
Description & Specifications

Specifications

Formula Weight 136.19
Chemical Name or Material 2,4-Dimethylbenzyl alcohol
Assay 97%
Molecular Formula C9H12O
CAS 16308-92-2
Boiling Point 120° to 121°C (13mmHg)
MDL Number MFCD00004624
Beilstein 1859679
Quantity 1g
Flash Point >110°C (230°F)