missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylbenzoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC160322500

 View more versions of this product

Catalog No. AC160322500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4-Dimethylbenzoic acid
Glass bottle
White
250g
97.5
98%
(CH3)2C6H3CO2H
09, 531
BKYWPNROPGQIFZ-UHFFFAOYSA-N
2,4-dimethylbenzoic acid
11897
150.18
98%
Authentic
267°C (727.0mmHg)
123°C to 127°C
611-01-8
100.0
C9H10O2
MFCD00002480
2,4 dimethylbenzoic acid
CC1=CC(=C(C=C1)C(=O)O)C
150.18
CHEBI:64811
Crystalline Powder

Chemical Identifiers

611-01-8
150.18
BKYWPNROPGQIFZ-UHFFFAOYSA-N
11897
2,4-dimethylbenzoic acid
C9H10O2
MFCD00002480
2,4 dimethylbenzoic acid
CHEBI:64811
CC1=CC(=C(C=C1)C(=O)O)C
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-246-2

SDS Documents