Please login to your online account to display your discounted pricing

2,4-Dimethylanisole, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  160001000

 View more versions of this product

Catalog No. AC160001000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 136.19
Quantity 100g
Color Clear colorless
Beilstein 06, 486
EINECS Number 229-794-9
TSCA TSCA
Density 0.9700g/mL
CAS Max % 100.0
Chemical Name or Material 2, 4-Dimethylanisole
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Refractive Index 1.5130 to 1.5150 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3OCH3
MDL Number MFCD00008377
CAS 6738-23-4
Synonym 1, 3-Dimethyl-4-methoxybenzene
CAS Min % 98.5

Specifications

Chemical Name or Material 2, 4-Dimethylanisole
Name Note 99%
CAS 6738-23-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3OCH3
Formula Weight 136.19
MDL Number MFCD00008377
Synonym 1, 3-Dimethyl-4-methoxybenzene