missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylanisole, 99%, ACROS Organics™

Chemical Identifiers

CAS 6738-23-4
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C9H12O
Linear Formula (CH3)2C6H3OCH3
Formula Weight 136.19
MDL Number MFCD00008377
Synonym 1, 3-Dimethyl-4-methoxybenzene
Chemical Name or Material 2, 4-Dimethylanisole, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC160001000 SDS Fisher Scientific Acros
160001000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6738-23-4
100.0
C9H12O
136.19
1, 3-Dimethyl-4-methoxybenzene
≥98.5%
98.5
99%
(CH3)2C6H3OCH3
MFCD00008377
2, 4-Dimethylanisole, 99%

Specifications

0.9700g/mL
100g
98.5
Authentic
1.5130 to 1.5150 (20°C, 589nm)
99%
(CH3)2C6H3OCH3
MFCD00008377
1, 3-Dimethyl-4-methoxybenzene
≥98.5%
Clear colorless
6738-23-4
100.0
Glass bottle
Liquid
C9H12O
136.19
06, 486
2, 4-Dimethylanisole, 99%

Safety and Handling

EINECSNumber : 229-794-9

TSCA : TSCA