Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dimethylaniline, 99%

Chemical Identifiers

Chemical Name or Material 2,4-Dimethylaniline
CAS 95-68-1
Molecular Formula C8H11N
Formula Weight 121.18
MDL Number MFCD00007738
UN Number UN1711
Beilstein 636243
Merck Index 14,10084
Synonym 2,4-Xylidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL027440P

  SDS
alfa aesar™
L027440P
2000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0274422

  SDS
alfa aesar™
L0274422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0274436

  SDS
alfa aesar™
L0274436
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.18
Chemical Name or Material 2,4-Dimethylaniline
UN Number UN1711
Health Hazard 2 GHS H Statement
H301-H311-H331-H351-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 99%
Molecular Formula C8H11N
MDL Number MFCD00007738
Beilstein 636243
Merck Index 14,10084
Synonym 2,4-Xylidine
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID
RTECS Number ZE8925000
Boiling Point 98° to 101°C (11mmHg)
Melting Point 16°C
Quantity 2000g
CAS 95-68-1
Flash Point 90°C (194°F)
Safety and Handling

Danger

GHS P Statement
P210-P260-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H351-H373-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.