Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate, 97%

Chemical Identifiers

Chemical Name or Material 2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate
CAS 64872-13-5
Formula Weight 378.81
MDL Number MFCD00274415
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55608MB

 
alfa aesar™
H55608MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55608MC

 
alfa aesar™
H55608MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 378.81
Chemical Name or Material 2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate
Physical Form Solid
Assay 97%
MDL Number MFCD00274415
CAS 64872-13-5
Quantity 25mg