Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide, ≥99%

Chemical Identifiers

Chemical Name or Material 2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide
CAS 3785-01-1
Formula Weight 380.27
MDL Number MFCD00011994
Beilstein 3783243
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5516303

  SDS
alfa aesar™
H5516303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55163MD

  SDS
alfa aesar™
H55163MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 380.27
Chemical Name or Material 2-[4-(Dimethylamino)styryl]-1-ethylpyridinium iodide
TSCA TSCA
Sensitivity Light Sensitive
Physical Form Solid
Assay ≥99%
MDL Number MFCD00011994
CAS 3785-01-1
Beilstein 3783243
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
RTECS Number UU3971000
Color Red
Melting Point 267°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C